2-(Trifluoromethoxy)phenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008427-1G 008427-5G
FLUO008427-1GEA 0 GBP
FLUO008427-1G FLUO008427-5G
2-(Trifluoromethoxy)phenyl isothiocyanate
2-(Trifluoromethoxy)phenyl isothiocyanate
Formula: C₈H₄F₃NOS
MW: 219.19 g/mol
MDL Number: MFCD00220860
CAS Number: 175205-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR