2,3,5-Trifluorophenyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009603-500MG
009603-1G
FLUO009603-500MGEA
0
GBP
FLUO009603-500MG
FLUO009603-1G
2,3,5-Trifluorophenyl isothiocyanate
2,3,5-Trifluorophenyl isothiocyanate
Formula:
C₇H₂F₃NS |
CAS Number:
362690-53-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...