2-Fluorophenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001385.5G 001385-25G 001385-10G 001385-50G
FLUO001385.5GEA 0 GBP
FLUO001385.5G FLUO001385-25G FLUO001385-10G FLUO001385-50G
2-Fluorophenyl isothiocyanate
2-Fluorophenyl isothiocyanate
Formula: C₇H₄FNS
MW: 153.18 g/mol
Boiling Pt: 54…58 °C (1 mmHg)
Melting Pt: 24…26 °C
Density: 1.24 g/cm³ (20 °C)
MDL Number: MFCD00004800
CAS Number: 38985-64-7
EINECS: 254-237-1
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR