1-Iodo-3-(trifluoromethoxy)benzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
013870-1G
013870-5G
013870-25G
FLUO013870-1GEA
0
GBP
FLUO013870-1G
FLUO013870-5G
FLUO013870-25G
1-Iodo-3-(trifluoromethoxy)benzene
1-Iodo-3-(trifluoromethoxy)benzene
Formula:
C₇H₄F₃IO MW: 288.01 g/mol |
MDL Number:
MFCD01090992 CAS Number: 198206-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...