1-Iodo-2-(trifluoromethoxy)benzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007557-100G 007557-25G
FLUO007557-100GEA 0 GBP
FLUO007557-100G FLUO007557-25G
1-Iodo-2-(trifluoromethoxy)benzene
1-Iodo-2-(trifluoromethoxy)benzene
Formula: C₇H₄F₃IO
MDL Number: MFCD00042410
CAS Number: 175278-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR