3-((Trifluoromethyl)thio)benzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
007998.25G
007998-5G
007998-1G
FLUO007998.25GEA
439
GBP
FLUO007998.25G
FLUO007998-5G
FLUO007998-1G
3-((Trifluoromethyl)thio)benzoic acid
3-((Trifluoromethyl)thio)benzoic acid
Formula:
C₈H₅F₃O₂S |
MDL Number:
MFCD00236337 CAS Number: 946-65-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...