5-Fluorosalicylic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001665-1G 001665-5G
FLUO001665-1GEA 0 GBP
FLUO001665-1G FLUO001665-5G
5-Fluorosalicylic acid
5-Fluorosalicylic acid
Formula: C₇H₅FO₃
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 180 °C
MDL Number: MFCD00002456
CAS Number: 345-16-4
EINECS: 206-455-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR