4-Fluoro-2-methoxybenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013328-1G 013328-5G
FLUO013328-1GEA 0 GBP
FLUO013328-1G FLUO013328-5G
4-Fluoro-2-methoxybenzoic acid
4-Fluoro-2-methoxybenzoic acid
Formula: C₈H₇FO₃
MW: 170.14 g/mol
MDL Number: MFCD00673010
CAS Number: 395-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR