3-Fluoro-4-methoxybenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005215-1G 005215-5G 005215-100G 005215-25G
FLUO005215-1GEA 0 GBP
FLUO005215-1G FLUO005215-5G FLUO005215-100G FLUO005215-25G
3-Fluoro-4-methoxybenzoic acid
3-Fluoro-4-methoxybenzoic acid
Formula: C₈H₇FO₃
Melting Pt: 211.5 °C
MDL Number: MFCD00060675
CAS Number: 403-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR