3-(Difluoromethoxy)benzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007853-1G 007853-5G
FLUO007853-1GEA 0 GBP
FLUO007853-1G FLUO007853-5G
3-(Difluoromethoxy)benzoic acid
3-(Difluoromethoxy)benzoic acid
Formula: C₈H₆F₂O₃
MDL Number: MFCD00236224
CAS Number: 4837-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR