2-Fluoro-6-methoxybenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF1122.1 019311-5G 019311-1G
CHMPCFF1122.1EA 0 GBP
CHMPCFF1122.1 FLUO019311-5G FLUO019311-1G
2-Fluoro-6-methoxybenzoic acid
2-Fluoro-6-methoxybenzoic acid
Formula: C₈H₇FO₃
MDL Number: MFCD00671764
CAS Number: 137654-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR