3-Fluoro-4-nitrobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

013031-5G 013031-1G 013031-25G
FLUO013031-5GEA 0 GBP
FLUO013031-5G FLUO013031-1G FLUO013031-25G
3-Fluoro-4-nitrobenzoic acid
3-Fluoro-4-nitrobenzoic acid
Formula: C₇H₄FNO₄
MDL Number: MFCD01862092
CAS Number: 403-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR