5-Amino-2-fluorobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005488-5G 005488-25G 005488-100G
FLUO005488-5GEA 0 GBP
FLUO005488-5G FLUO005488-25G FLUO005488-100G
5-Amino-2-fluorobenzoic acid
5-Amino-2-fluorobenzoic acid
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
MDL Number: MFCD00077449
CAS Number: 56741-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR