Ethyl-4-fluorobenzoate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Fluorobenzoic acid ethyl ester, Ethyl 4-fluorobenzoate

006108-25G 006108-100G 006108-250G 006108-50G
FLUO006108-25GEA 0 GBP
FLUO006108-25G FLUO006108-100G FLUO006108-250G FLUO006108-50G
Ethyl-4-fluorobenzoate
Ethyl 4-fluorobenzoate
Formula: C₉H₉FO₂
MW: 168.17 g/mol
Boiling Pt: 210
Melting Pt: 26
Density: 1.146 g/cm³ (20 °C)
MDL Number: MFCD00000351
CAS Number: 451-46-7
EINECS: 207-194-8
Merck Index: 13,04198

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR