Methyl-3-fluorobenzoate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Fluorobenzoic acid emthyl ester, Methyl 3-fluorobenzoate
009502-25G
009502-100G
FLUO009502-25GEA
0
GBP
FLUO009502-25G
FLUO009502-100G
Methyl-3-fluorobenzoate
Methyl 3-fluorobenzoate
Formula:
C₈H₇FO₂ MW: 154.14 g/mol |
MDL Number:
MFCD03094302 CAS Number: 455-68-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...