Ethyl-2-fluorobenzoate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006106-25G 006106-100G 006106-250G
FLUO006106-25GEA 0 GBP
FLUO006106-25G FLUO006106-100G FLUO006106-250G
Ethyl-2-fluorobenzoate
Ethyl 2-fluorobenzoate
Formula: C₉H₉FO₂
MDL Number: MFCD00039214
CAS Number: 443-26-5
EINECS: 207-134-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR