4-Fluorobenzhydrazide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Fluorobenzoic hydrazide, 4-Fluorobenzene-1-carbohydrazide, 4-Fluorobenzoic acid hydrazide, 4-Fluorobenzohydrazide

006190-5G 006190-25G
FLUO006190-5GEA 0 GBP
FLUO006190-5G FLUO006190-25G
4-Fluorobenzhydrazide
4-Fluorobenzhydrazide
Formula: C₇H₇FN₂O
MDL Number: MFCD00060562
CAS Number: 456-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR