3-Fluorobenzhydrazide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Fluorobenzene-1-carbohydrazide, 3-Fluorobenzoic acid hydrazide, 3-Fluorobenzoic hydrazide, 3-Fluorobenzohydrazide

006189-5G 006189-1G 006189-25G
FLUO006189-5GEA 0 GBP
FLUO006189-5G FLUO006189-1G FLUO006189-25G
3-Fluorobenzhydrazide
3-Fluorobenzhydrazide
Formula: C₇H₇FN₂O
MDL Number: MFCD00060561
CAS Number: 499-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR