4-((Trifluoromethyl)thio)benzoyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005859-5G 005859-1G 005859-25G
FLUO005859-5GEA 0 GBP
FLUO005859-5G FLUO005859-1G FLUO005859-25G
4-((Trifluoromethyl)thio)benzoyl chloride
4-((Trifluoromethyl)thio)benzoyl chloride
Formula: C₈H₄ClF₃OS
Boiling Pt: 96 °C (18 mmHg)
Density: 1.43 g/cm³ (20 °C)
MDL Number: MFCD01631632
CAS Number: 330-14-3
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR