4-(Trifluoromethoxy)benzoyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
007548-1G
007548-5G
007548-25G
FLUO007548-1GEA
0
GBP
FLUO007548-1G
FLUO007548-5G
FLUO007548-25G
4-(Trifluoromethoxy)benzoyl chloride
4-(Trifluoromethoxy)benzoyl chloride
Formula:
C₈H₄ClF₃O₂ Boiling Pt: 90…92 °C (15 mmHg) Density: 1.42 g/cm³ (20 °C) |
MDL Number:
MFCD00052329 CAS Number: 36823-88-8 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...