Pentafluorobenzoyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2,3,4,5,6-Pentafluorobenzoyl chloride, Pentafluorobenzoylchloride, PFBC

001351-25G 001351-100G
FLUO001351-25GEA 0 GBP
FLUO001351-25G FLUO001351-100G
Pentafluorobenzoyl chloride
Pentafluorobenzoyl chloride
Formula: C₇ClF₅O
MW: 230.52 g/mol
Boiling Pt: 158
Density: 1.6 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000657
CAS Number: 2251-50-5
EINECS: 218-842-4
UN: 3265

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR