4-Fluorobenzoyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

001636-100G 001636-500G 001636-25G
FLUO001636-100GEA 0 GBP
FLUO001636-100G FLUO001636-500G FLUO001636-25G
4-Fluorobenzoyl chloride
4-Fluorobenzoyl chloride
Formula: C₇H₄ClFO
Boiling Pt: 180 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000684
CAS Number: 403-43-0
EINECS: 206-961-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR