3,4-Difluorobenzoyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

002083-25G 002083-100G
FLUO002083-25GEA 0 GBP
FLUO002083-25G FLUO002083-100G
3,4-Difluorobenzoyl chloride
3,4-Difluorobenzoyl chloride
Formula: C₇H₃ClF₂O
Boiling Pt: 61 °C (1013 hPa)
Density: 1.41 g/cm³ (20 °C)
MDL Number: MFCD00009930
CAS Number: 76903-88-3
EINECS: 278-570-7
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR