2-Fluorobenzoyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: o-Fluorobenzoyl chloride

001420-100G 001420-500G
FLUO001420-100GEA 0 GBP
FLUO001420-100G FLUO001420-500G
2-Fluorobenzoyl chloride
2-Fluorobenzoyl chloride
Formula: C₇H₄ClFO
Boiling Pt: 206 °C (1013 hPa)
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000656
CAS Number: 393-52-2
EINECS: 206-887-2
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR