2,5-Difluorobenzoyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

001937-5G 001937-25G
FLUO001937-5GEA 0 GBP
FLUO001937-5G FLUO001937-25G
2,5-Difluorobenzoyl chloride
2,5-Difluorobenzoyl chloride
Formula: C₇H₃ClF₂O
Boiling Pt: 92…93 °C (34 mmHg)
Density: 1.42 g/cm³ (20 °C)
MDL Number: MFCD00009929
CAS Number: 35730-09-7
EINECS: 252-701-8
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR