2,4,6-Trifluorobenzoyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

007496-5G 007496-1G 007496-25G
FLUO007496-5GEA 0 GBP
FLUO007496-5G FLUO007496-1G FLUO007496-25G
2,4,6-Trifluorobenzoyl chloride
2,4,6-Trifluorobenzoyl chloride
Formula: C₇H₂ClF₃O
Boiling Pt: 186 °C (1013 hPa)
Melting Pt: 41 °C
MDL Number: MFCD00070810
CAS Number: 79538-29-7
UN: 1760
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR