2,3-Difluorobenzoyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
004788-5G
004788-25G
FLUO004788-5GEA
0
GBP
FLUO004788-5G
FLUO004788-25G
2,3-Difluorobenzoyl chloride
2,3-Difluorobenzoyl chloride
Formula:
C₇H₃ClF₂O Boiling Pt: 85…87 °C (1013 hPa) Density: 1.423 g/cm³ (20 °C) |
MDL Number:
MFCD00042499 CAS Number: 18355-73-2 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...