2,3,6-Trifluorobenzoyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
007495-5G
007495-1G
FLUO007495-5GEA
0
GBP
FLUO007495-5G
FLUO007495-1G
2,3,6-Trifluorobenzoyl chloride
2,3,6-Trifluorobenzoyl chloride
Formula:
C₇H₂ClF₃O Boiling Pt: 77…79 °C (20 mmHg) Density: 1.48 g/cm³ (20 °C) |
MDL Number:
MFCD00061203 CAS Number: 189807-20-3 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...