4-(Trifluoromethoxy)benzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001405-5G
001405-25G
001405-100G
FLUO001405-5GEA
0
GBP
FLUO001405-5G
FLUO001405-25G
FLUO001405-100G
4-(Trifluoromethoxy)benzonitrile
4-(Trifluoromethoxy)benzonitrile
Formula:
C₈H₄F₃NO Boiling Pt: 192 °C (1013 hPa) Density: 1.284 g/cm³ (20 °C) |
MDL Number:
MFCD00039474 CAS Number: 332-25-2 EINECS: 206-363-3 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...