2-Fluorobenzonitrile, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ200
196649-25G
196649-5G
SIAL196649-25GEA
50.9
GBP
SIAL196649-25G
SIAL196649-5G
2-Fluorobenzonitrile, Sigma-Aldrich®
2-Fluorobenzonitrile
Formula:
C₇H₄FN Boiling Pt: 190 °C (1013 hPa) Melting Pt: 35…37 °C Density: 1.11 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001773 CAS Number: 394-47-8 EINECS: 206-897-7 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...