2,3,5-Trifluorobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007957-5G 007957-10G 007957-1G
FLUO007957-5GEA 0 GBP
FLUO007957-5G FLUO007957-10G FLUO007957-1G
2,3,5-Trifluorobenzonitrile
2,3,5-Trifluorobenzonitrile
Formula: C₇H₂F₃N
MW: 157.09 g/mol
MDL Number: MFCD00083539
CAS Number: 241154-09-6
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR