4-(Trifluoromethoxy)benzaldehyde
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001404-5G
001404-25G
001404-100G
FLUO001404-5GEA
0
GBP
FLUO001404-5G
FLUO001404-25G
FLUO001404-100G
4-(Trifluoromethoxy)benzaldehyde
4-(Trifluoromethoxy)benzaldehyde
Formula:
C₈H₅F₃O₂ MW: 190.12 g/mol Boiling Pt: 80 Density: 1.331 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00041530 CAS Number: 659-28-9 EINECS: 211-531-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...