4-(Difluoromethoxy)benzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004830-5G 004830-25G
FLUO004830-5GEA 0 GBP
FLUO004830-5G FLUO004830-25G
4-(Difluoromethoxy)benzaldehyde
4-(Difluoromethoxy)benzaldehyde
Formula: C₈H₆F₂O₂
MDL Number: MFCD00042252
CAS Number: 73960-07-3
EINECS: 277-653-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR