3-(Trifluoromethoxy)benzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001541-5G 001541-25G
FLUO001541-5GEA 0 GBP
FLUO001541-5G FLUO001541-25G
3-(Trifluoromethoxy)benzaldehyde
3-(Trifluoromethoxy)benzaldehyde
Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Boiling Pt: 83…86 °C (24 mmHg)
Melting Pt: 450
Density: 1.33 g/cm³ (20 °C)
MDL Number: MFCD00042406
CAS Number: 52771-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR