2,3,4-Trifluorobenzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

007476-5G 007476-1G
FLUO007476-5GEA 0 GBP
FLUO007476-5G FLUO007476-1G
2,3,4-Trifluorobenzaldehyde
2,3,4-Trifluorobenzaldehyde
Formula: C₇H₃F₃O
MW: 160.1 g/mol
Boiling Pt: 77…79 °C (20 mmHg)
Density: 1,404 g/cm³ (20 °C)
MDL Number: MFCD00061230
CAS Number: 161793-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR