4-(Trifluoromethoxy)phenylboronic acid ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
B23233.03
B23233.06
B23233.03EA
22.8
GBP
B23233.03
B23233.06
4-(Trifluoromethoxy)phenylboronic acid ≥98%
4-(Trifluoromethoxy)phenylboronic acid
Purity: 98%
CAS No.: 139301-27-2
Molecular Formula: C7H6BF3O3
Formula Weight: 205.93
Melting Point: ca 120-125°
Beilstein Reference: 8548201
Formula:
C₇H₆BF₃O₃ MW: 205.93 g/mol Melting Pt: ∼ 120…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074648 CAS Number: 139301-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...