2-(Trifluoromethoxy)phenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013043-1G 013043-5G
FLUO013043-1GEA 0 GBP
FLUO013043-1G FLUO013043-5G
2-(Trifluoromethoxy)phenylboronic acid
2-(Trifluoromethoxy)phenylboronic acid
Formula: C₇H₆BF₃O₃
MDL Number: MFCD01320763
CAS Number: 175676-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR