3,5-Difluorophenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

013044-1G 013044-25G 013044-5G
FLUO013044-1GEA 0 GBP
FLUO013044-1G FLUO013044-25G FLUO013044-5G
3,5-Difluorophenylboronic acid
3,5-Difluorophenylboronic acid
Formula: C₆H₅BF₂O₂
MDL Number: MFCD01318138
CAS Number: 156545-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR