3,4-Difluorophenylboronic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3,4-Difluorobenzeneboronic acid, (3,4-Difluorophenyl)metaboric acid, (3,4-Difluorophenyl)boronic acid, Dihydroxy(3,4-difluorophenyl)borane
M-Clarity™ quality level = MQ100
465089-25G
465089-5G
SIAL465089-25GEA
168
GBP
SIAL465089-25G
SIAL465089-5G
3,4-Difluorophenylboronic acid, Sigma-Aldrich®
3,4-Difluorophenylboronic acid
Formula:
C₆H₅BF₂O₂ MW: 157.91 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00807405 CAS Number: 168267-41-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...