tert-Butyl perbenzoate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: tert-Butyl peroxybenzoate, Luperox(R) P, tert-Butyl peroxybenzoate

M-Clarity™ quality level = MQ200

159042-100G 159042-500G 159042-5G
SIAL159042-100GEA 56.2 GBP
SIAL159042-100G SIAL159042-500G SIAL159042-5G
tert-Butyl perbenzoate, Sigma-Aldrich®
tert-Butyl perbenzoate
Formula: C₁₁H₁₄O₃
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 8 °C
Density: 1.043 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00008802
CAS Number: 614-45-9
EINECS: 210-382-2
UN: 3103
ADR: 5.2,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR