Terephthalic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

185361-100G 185361-500G 185361-5G
SIAL185361-100GEA 59.5 GBP
SIAL185361-100G SIAL185361-500G SIAL185361-5G
Terephthalic acid, Sigma-Aldrich®
Terephthalic acid
Formula: C₈H₆O₄
MW: 166.13 g/mol
Boiling Pt: 392
Density: 1.472 g/cm³ (20 °C)
Flash Pt: 260
Storage Temperature: Ambient
MDL Number: MFCD00002558
CAS Number: 100-21-0
EINECS: 202-830-0
Merck Index: 12,09306

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR