Phthalic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Pathalic acid

80010-100G 80010-1KG
SIAL80010-100GEA 53.3 GBP
SIAL80010-100G SIAL80010-1KG
Phthalic acid, Sigma-Aldrich®
Phthalic acid
Formula: C₈H₆O₄
MW: 166.13 g/mol
Boiling Pt: 315…317 °C (1013 hPa)
Melting Pt: 79 °C
Density: 1.576 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002467
CAS Number: 88-99-3
EINECS: 201-873-2
Merck Index: 12,07527

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR