Phthalic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Pathalic acid
M-Clarity™ quality level = MQ200
402915-1KG
402915-250G
402915-25G
SIAL402915-1KGEA
145
GBP
SIAL402915-1KG
SIAL402915-250G
SIAL402915-25G
Phthalic acid, Sigma-Aldrich®
Phthalic acid
Formula:
C₈H₆O₄ MW: 166.13 g/mol Boiling Pt: 315…317 °C (1013 hPa) Melting Pt: 79 °C Density: 1.576 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002467 CAS Number: 88-99-3 EINECS: 201-873-2 Merck Index: 12,07527 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...