Sodium salicylate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

71945-250G 71945-1KG
SIAL71945-250GEA 77.2 GBP
SIAL71945-250G SIAL71945-1KG
Sodium salicylate, Sigma-Aldrich®
Sodium salicylate
Formula: HOC₆H₄COONa
Melting Pt: 200 °C
Density: 0.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002440
CAS Number: 54-21-7
EINECS: 200-198-0
Merck Index: 12,08819

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR