Sodium salicylate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Sodium 2-hydroxybenzoate, 2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt
MQ100
31493-1KG
31493-250G
SIAL31493-1KGEA
192
GBP
SIAL31493-1KG
SIAL31493-250G
Sodium salicylate, Sigma-Aldrich®
Sodium salicylate
Formula:
HOC₆H₄COONa MW: 160.10 g/mol Melting Pt: 200 Density: 0.32 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002440 CAS Number: 54-21-7 EINECS: 200-198-0 Merck Index: 12,08819 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...