4-Hydroxybenzoic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
p-Hydroxybenzoic Acid
M-Clarity™ quality level = MQ300
W398608-1KG
W398608-5KG
SIALW398608-1KGEA
84.6
GBP
SIALW398608-1KG
SIALW398608-5KG
4-Hydroxybenzoic acid, Sigma-Aldrich®
4-Hydroxybenzoic acid
Formula:
C₇H₆O₃ MW: 138.12 g/mol Boiling Pt: 336 °C (1013 hPa) Melting Pt: 210…211 °C Density: 1.398 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002547 CAS Number: 99-96-7 EINECS: 202-804-9 Merck Index: 13,04837 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...