4-Hydroxybenzoic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ200
H20059-100G
H20059-1KG
H20059-500G
SIALH20059-100GEA
43
GBP
SIALH20059-100G
SIALH20059-1KG
SIALH20059-500G
4-Hydroxybenzoic acid, Sigma-Aldrich®
4-Hydroxybenzoic acid
Formula:
C₇H₆O₃ Boiling Pt: 336 °C (1013 hPa) Melting Pt: 210…211 °C Density: 1.398 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002547 CAS Number: 99-96-7 EINECS: 202-804-9 Merck Index: 13,04837 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...