Syringic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Hydroxy-3,5-dimethoxybenzoic acid
M-Clarity™ quality level = MQ100
S6881-100G
S6881-10G
S6881-25G
S6881-5G
SIALS6881-100GEA
1110
GBP
SIALS6881-100G
SIALS6881-10G
SIALS6881-25G
SIALS6881-5G
Syringic acid, Sigma-Aldrich®
Syringic acid
Formula:
C₉H₁₀O₅ MW: 198.18 g/mol Boiling Pt: 351 °C (1013 hPa) Melting Pt: 204…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002552 CAS Number: 530-57-4 EINECS: 208-486-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...