4-Nitrosalicylic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Hydroxy-4-nitrobenzoic acid
012256-25G
012256-1G
012256-5G
FLUO012256-25GEA
0
GBP
FLUO012256-25G
FLUO012256-1G
FLUO012256-5G
4-Nitrosalicylic acid
4-Nitrosalicylic acid
Formula:
C₇H₅NO₅ MW: 183.12 g/mol |
MDL Number:
MFCD00071505 CAS Number: 619-19-2 EINECS: 210-584-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...