4-Nitrobenzoic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ200

461091-1KG 461091-250G
SIAL461091-1KGEA 95.5 GBP
SIAL461091-1KG SIAL461091-250G
4-Nitrobenzoic acid, Sigma-Aldrich®
4-Nitrobenzoic acid
Formula: C₇H₅NO₄
Boiling Pt: 359 °C (1013 hPa)
Melting Pt: 121 °C
Density: 1.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007352
CAS Number: 62-23-7
EINECS: 200-526-2
Merck Index: 12,06686

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR